Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxO1/FKHR Antibody, Novus Biologicals™
SDP

Catalog No. NB1002312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002312 Supplier Novus Biologicals Supplier No. NB1002312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen FoxO1/FKHR
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000- 1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q12778
Gene Alias FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Gene Symbols FOXO1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 600 and the C-terminus (residue 652) of human Forkhead Box O1a using the numbering given in Swiss-Prot entry Q12778 (GeneID 2308).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308
Test Specificity Expected to cross react based on 100% sequence identity: Panda, Orangutan, Rhesus Monkey, Chimpanzee, Northern white-cheeked gibbon, Gray short-tailed opossum, Crab-eating macaque, African elephant, Small-eared galago, Chinese hamster, Little brown bat, White-tufted-ear marmoset.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.