Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO1A/3/4 Antibody (4E7), Novus Biologicals™
SDP

Catalog No. NBP141196 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141196 Supplier Novus Biologicals Supplier No. NBP141196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FOXO1A/3/4 Monoclonal specifically detects FOXO1A/3/4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FOXO1A/3/4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 40000000
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-2 ug/ml, Immunohistochemistry 1:10 - 1:500
Gene Alias FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Gene Symbols FOXO1
Host Species Mouse
Immunogen Amino acids 242-655 of FKHR expressed in E. coli.
Molecular Weight of Antigen 70 kDa
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2308
Test Specificity This antibody will recognize human FKHR.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.