Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO3 Rabbit anti-Human, Mouse, Clone: 8B9X5, Novus Biologicals™
SDP

Catalog No. NB168693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168693 100 μg
NB168694 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168693 Supplier Novus Biologicals Supplier No. NBP316801100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FOXO3 Monoclonal antibody specifically detects FOXO3 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen FOXO3
Applications Western Blot
Classification Monoclonal
Clone 8B9X5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 574-673 of human FOXO3 (O43524).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2309
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.