Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXO3 Rabbit anti-Human, Mouse, Clone: 8B9X5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | FOXO3 |
---|---|
Clone | 8B9X5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FOXO3 Monoclonal antibody specifically detects FOXO3 in Human, Mouse samples. It is validated for Western BlotSpecifications
FOXO3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, DNA Repair, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2309 | |
IgG | |
Affinity purified |
8B9X5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739 | |
A synthetic peptide corresponding to a sequence within amino acids 574-673 of human FOXO3 (O43524). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title