Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO3a Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140201
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7659 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7659 Supplier Bethyl Laboratories, Inc Supplier No. A300454AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.

FoxO (forkhead box O) proteins are a family of transcription factors characterized by the distinct forkhead domain. The FoxO proteins act in the insulin signaling pathway, but FoxO3 has also been shown to play a role in the regulation of apoptosis, cell cycle progression and cancer. Several splice variants of FoxO3 have been reported.

Specifications

Antigen FOXO3a
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FOXO3
Gene Accession No. O43524, Q9WVH4
Gene Alias FkhR
Gene Symbols FOXO3
Host Species Rabbit
Immunogen Between 625 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2309, 56484
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.