Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO4 Antibody (OTI1A4), HRP, Novus Biologicals™
SDP

Catalog No. NBP271198H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271198H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271198H Supplier Novus Biologicals Supplier No. NBP271198H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FOXO4 Monoclonal specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FOXO4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A4
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7
Gene Symbols FOXO4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 206-505 of human FOXO4 (NP_005929) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4303.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.