Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP1 Antibody (837016), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB303533 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303533 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303533 Supplier Novus Biologicals Supplier No. FAB45341N

Mouse Monoclonal Antibody

FoxP1 Monoclonal antibody specifically detects FoxP1 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen FoxP1
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 837016
Conjugate Alexa Fluor 700
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4
Host Species Mouse
Immunogen E. coli-derived recombinant human FoxP1, Lys548-Glu677, Accession # Q9H334
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 27086
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.