Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP1 Antibody (JC12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB10065125I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065125I 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065125I Supplier Novus Biologicals Supplier No. NB10065125IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxP1 Monoclonal specifically detects FoxP1 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen FoxP1
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone JC12
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4
Gene Symbols FOXP1
Host Species Mouse
Immunogen Human FOXP1 [Uniprot# Q9H334]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27086
Test Specificity Does not recognize closely related molecules FOXP2, FOXP3 or FOXP4.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.