Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP1 Antibody (rFOXP1/6902), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB187020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187020 Supplier Novus Biologicals Supplier No. NBP314022IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FoxP1 Monoclonal antibody specifically detects FoxP1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen FoxP1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rFOXP1/6902
Conjugate DyLight 755
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4
Host Species Mouse
Immunogen Recombinant fragment (around aa350-450) of human FoxP1 protein (exact sequence is proprietary) (Uniprot: Q9H334)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 27086
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.