Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXP1 Monoclonal Antibody (1G1), Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 61309

Description
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
FOXP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
P58462 | |
FOXP1 | |
This FOXP1 antibody was raised against a recombinant protein corresponding to amino acids 495-578 of mouse FOXP1. This antibody detects three isoforms. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
ChIP Assay, Gel Shift, Western Blot | |
1G1 | |
Unconjugated | |
FOXP1 | |
12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; fork head-related protein like B; forkhead box P1; Forkhead box protein P1; Forkhead-related transcription factor 1; FOXP1; glutamine-rich factor 1; hFKH1B; HSPC215; Mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1 | |
Mouse | |
Protein A | |
RUO | |
108655 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction