Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXP1 Monoclonal Antibody (1G1), Active Motif™

Mouse Monoclonal Antibody
$132.55 - $397.65
Specifications
Antigen | FOXP1 |
---|---|
Clone | 1G1 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, Gel Shift, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3321
![]() ![]() |
Active Motif
61310 |
10 μg |
Each for $132.55
|
|
|||||
50-199-3324
![]() ![]() |
Active Motif
61309 |
100 μg |
Each for $397.65
|
|
|||||
Description
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
FOXP1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; fork head-related protein like B; forkhead box P1; Forkhead box protein P1; Forkhead-related transcription factor 1; FOXP1; glutamine-rich factor 1; hFKH1B; HSPC215; Mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1 | |
FOXP1 | |
IgG1 | |
Protein A | |
Antibody |
1G1 | |
ChIP Assay, Gel Shift, Western Blot | |
Unconjugated | |
Mouse | |
Mouse | |
P58462 | |
108655 | |
This FOXP1 antibody was raised against a recombinant protein corresponding to amino acids 495-578 of mouse FOXP1. This antibody detects three isoforms. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
FOXP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title