Learn More
FoxP3 Antibody (16J4G6) - Delta 2 (exon 2 deleted) specific, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | FOXP3 Delta 2 (exon 2 deleted) specific |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 16J4G6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin |
| Gene Accession No. | Q9BZS1 |
| Gene Alias | AIID, DIETER, Forkhead Box P3, Forkhead Box Protein P3, FOXP3, FOXP3 Delta, FOXP3 Delta2 (exon 2 deleted) specific, FOXP3Delta2, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, PIDX, scurfin, XPID |
| Gene Symbols | FOXP3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.