Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody (376209R), HRP, Novus Biologicals™
SDP

Catalog No. NB345912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345912 Supplier Novus Biologicals Supplier No. FAB8970H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FoxP3
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 376209R
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Host Species Mouse
Immunogen E.coli-derived recombinant human FoxP3, Gln105-Lys200, Accession # Q9BZS1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 50943
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.