Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10039002Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10039002Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10039002Z Supplier Novus Biologicals Supplier No. NB10039002AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FoxP3 Polyclonal specifically detects FoxP3 in Bovine, Human, Mouse, Rat samples. It is validated for Dual RNAScope ISH-IHC, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot.

Specifications

Antigen FoxP3
Applications Dual RNAScope ISH-IHC, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Gene Symbols FOXP3
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot# Q99JB6]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 50943
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.