Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10039002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10039002 0.1 mL
NB10039002T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10039002 Supplier Novus Biologicals Supplier No. NB10039002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 24 publications

FoxP3 Polyclonal specifically detects FoxP3 in Bovine, Human, Mouse, Rat samples. It is validated for Dual RNAScope ISH-IHC, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Multiplex Immunoassay, Single Cell Western, Western Blot.

Specifications

Antigen FoxP3
Applications Dual RNAScope ISH-IHC, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Multiplex, Single Cell Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:8000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 26485285), Single Cell Western 100 ug/mL, Dual RNAscope ISH-IHC
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q99JB6
Gene Alias AIID, AIIDMGC141961, anti-human foxp3, anti-mouse foxp3, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 ihc, foxp3 paraffin, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Gene Symbols FOXP3
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot# Q99JB6]
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity Mouse. This antibody shows reactivity against mouse CD4+CD25+T cells. There appears to be no reactivity against human Jurkat and mouse CD4+CD25-T cell lysates. Human reactivity reported in scientific literature (PMID: 23922187).
Target Species Bovine, Mouse, Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.