Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 24 publications

$254.00 - $594.00

Specifications

Antigen FoxP3
Concentration 1.0 mg/mL
Dilution Western Blot 1:2000, Flow Cytometry 1:8000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 26485285), Single Cell Western 100 ug/mL, Dual RNAscope ISH-IHC
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10039002T
SDP
View Documents
Novus Biologicals
NB10039002SS
0.025 mL
Each for $254.00
Only null left
Add to Cart
 
NB10039002
SDP
View Documents
Novus Biologicals
NB10039002
0.1 mL
Each for $594.00
Only null left
Add to Cart
 
Description

Description

FoxP3 Polyclonal specifically detects FoxP3 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Single Cell Western, Dual RNAscope ISH-IHC.
Specifications

Specifications

FoxP3
Western Blot 1:2000, Flow Cytometry 1:8000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 26485285), Single Cell Western 100 ug/mL, Dual RNAscope ISH-IHC
Polyclonal
Purified
RUO
Human, Mouse, Rat
Q99JB6
50943
A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot# Q99JB6]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators
PBS with 0.05% Sodium Azide
AIID, AIIDMGC141961, anti-human foxp3, anti-mouse foxp3, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 ihc, foxp3 paraffin, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
FOXP3
IgG
Epitope affinity purified
Mouse. This antibody shows reactivity against mouse CD4+CD25+T cells. There appears to be no reactivity against human Jurkat and mouse CD4+CD25-T cell lysates. Human reactivity reported in scientific literature (PMID: 23922187).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.