Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB600246C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600246C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600246C Supplier Novus Biologicals Supplier No. NB600246C

Rabbit Polyclonal Antibody

FoxP3 Polyclonal antibody specifically detects FoxP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen FoxP3
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9BZS1
Gene Alias AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Gene Symbols FOXP3
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal region of human FOXP3 (between residues 1-50). [UniProt# Q9BZS1]
Molecular Weight of Antigen 47.2 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 50943
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.