Learn More
Description
Specifications
Specifications
| Antigen | FoxP3 |
| Applications | Dual RNAScope ISH-IHC, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting |
| Formulation | PBS with No Preservative |
| Gene Alias | AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked |
| Gene Symbols | FOXP3 |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot# Q99JB6] |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
