Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Mouse anti-Human , BB700, Clone: 236A/E7, BD Biosciences™
Mouse Monoclonal Antibody
$215.00 - $444.00
Specifications
Antigen | FoxP3 |
---|---|
Clone | 236A/E7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Flow CytometrySpecifications
FoxP3 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human full-length FoxP3 Recombinant Protein | |
Primary |
236A/E7 | |
Monoclonal | |
Mouse | |
Human | |
Forkhead box protein P3; Scurfin; AIID; IPEX; JM2; DIETER; PIDX; XPID | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB700 under optimum conditions, and unconjugated antibody and free BD Horizon BB700 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title