Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Mouse anti-Human, PE, Clone: 259D/C7, BD

Mouse Monoclonal Antibody

$212.00 - $430.50

Specifications

Antigen FoxP3
Clone 259D/C7
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB560082
View Documents Promotion Details
BD Biosciences
560082
25 Tests
Each for $212.00
Only null left
Add to Cart
 
BDB560046
View Documents Promotion Details
BD Biosciences
560046
100 Tests
Each for $430.50
Only null left
Add to Cart
 
Description

Description

The 259D/C7 antibody reacts with the human FoxP3 transcription factor, a member of the forkhead or winged helix family of transcription factors. The expression of FoxP3, also known as Scurfin, IPEX and JM2, has been found to be associated with CD4+ regulatory T cells and represents a specific marker for these cells. Flow-cytometric analysis has shown that FoxP3 is expressed by the majority of CD4+CD25+high T cells in peripheral blood while less than half of CD4+CD25int cell population are FoxP3 positive. Approximately 5-10% of peripheral CD4+ cells are CD4+CD25+ T regulatory cells. T regulatory cells are thought to play a critical role in the control of T cell mediated autoimmunity by suppressing the proliferation and cytokine production of other T cells. To support this hypothesis, it has been found that FOXP3 is mutated in scurfy (sf) mice. The 259D/C7 antibody reacts with all currently identified isoforms of human FoxP3 and is cross-reactive with Cynomolgus, Rhesus and Baboon.

Specifications

Specifications

FoxP3
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1
Affinity Purified
Scurfin, IPEX, JM2
259D/C7
Monoclonal
Mouse
Baboon, Cynomolgus, Human, Rhesus Monkey
FoxP3 recombinant protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.