Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056473 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056473 0.1 mg
NB10056473T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056473 Supplier Novus Biologicals Supplier No. NB10056473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FPR1 Polyclonal specifically detects FPR1 in Human, Mouse, Primate samples. It is validated for Simple Western, Western Blot.

Specifications

Antigen FPR1
Applications Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Simple Western 100 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P21462
Gene Alias fMet-Leu-Phe receptor, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor
Gene Symbols FPR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 176-190 of human FPR1 (CTFNFSPWTNDPKER) was used as the immunogen
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2357
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimp, gorilla and orangutan, and 67% homologous in mouse.
Target Species Human, Mouse, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.