Learn More
Description
Specifications
Specifications
| Antigen | FPRL1/FPR2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24086560), Electron Microscopy |
| Formulation | PBS, 30% Glycerol with 0.01% Sodium Azide |
| Gene Accession No. | P25090 |
| Gene Alias | ALXR, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4RFMLP-R-I, N-formyl peptide receptor 2 |
| Gene Symbols | FPR2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
