Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPRL2/FPR3 Antibody (374822), Biotin, Novus Biologicals™
SDP

Catalog No. NB371291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371291 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371291 Supplier Novus Biologicals Supplier No. FAB3896B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen FPRL2/FPR3
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 374822
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FPRL2, Met1-Met352, Accession # P25089
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2359
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.