Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPRL2/FPR3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226058 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226058 Supplier Novus Biologicals Supplier No. NBP226058
Add to Cart
Add to Cart

Goat Polyclonal Antibody

FPRL2/FPR3 Polyclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen FPRL2/FPR3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Gene Symbols FPR3
Host Species Goat
Immunogen Peptide with sequence C-DSAQTSNTDTTS corresponding to C-Terminus according to NP_002021.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2359
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.