Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FR 171113

Click to view available options
Quantity:
10 mg
50 mg
Description
FR 171113 is a protease-activated receptor 1 (PAR1) antagonist. Exhibits potent antiplatelet activity in vitro; inhibits thrombin TRAP-6-induced platelet aggregation (IC50 = 2.5 μM) with no effect on coagulation time.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | FR 171113 |
| Purity | >98% |
| Molecular Weight (g/mol) | 469.73 |
| Inhibitors | FR 171113 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction