Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR Men's FR/CP Lab Coat

Catalog No. 19800051 Shop All Red Kap Industries Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Long
Regular
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides dual hazard protection with flame resistance and chemical protection. Bulwark™ FR FR/CP Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldCXP™ to repel small amounts of liquid chemicals.

Provides dual hazard protection with flame resistance and chemical protection. Bulwark™ FR Lab Coats are lightweight, breathable, and comfortable to wear. They employ Nomex™ IIIA fabric to prevent ignition and ShieldCXP™ to repel small amounts of liquid chemicals.

  • Nomex™ IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
  • Nomex™ knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
  • Black lay-down collar
  • Concealed snap front closure
  • Splash-resistant side pass-through for easy inner pocket access
  • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
  • Available in a variety of sizes with regular and long variants

Certifications

NFPA 2112 UL-Certified

Compliance

Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire. UL certified.

Specifications

Size Large
Gender Men's
Material Nomex IIIA
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Style Regular
No. of Pockets 3
Finish Westex ShieldCXP Chemical Splash Protection
Size (English) Chest 43.25 in.
Size (Metric) Chest 109.9 cm
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9cm
Weight (English) 4.5 oz.
Weight (Metric) 128 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.