Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FR4 Monoclonal Antibody (eBioTH6), Functional Grade, eBioscience™

Catalog No. p-7173187
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-145-64 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-145-64 Supplier Invitrogen™ Supplier No. 16544685
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody eBioTh6 recognizes FR4, also known as FR delta and FBP3 (folate binding protein3). FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naive T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors. Based on co-staining studies, the epitopes recognized by eBioTH6 and eBio12A5 are different thereby allowing functional studies with TH6 to be evaluated with eBio12A5. Applications Reported:This eBioTH6 antibody has been reported for use in flow cytometric analysis, immunoprecipitation and in vivo depletion. Applications Tested: This eBioTH6 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greate...

FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naive T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FR4
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone eBioTH6
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Izumo1r
Gene Accession No. Q9EQF4
Gene Alias 0910001L11Rik; C86255; Folate receptor 4; folate receptor 4 (delta); folate receptor 4 (delta) homolog; folate receptor 4, delta (putative); folate receptor delta; Folate-binding protein 3; Folbp3; Folr4; FR-delta; IZUMO1 receptor protein JUNO; IZUMO1 receptor, JUNO; Izumo1r; Juno; probable folate receptor delta; sperm-egg fusion protein Juno
Gene Symbols Izumo1r
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64931
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.