Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRA-1/FOSL1 Antibody (OTI12F9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271351B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271351B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271351B Supplier Novus Biologicals Supplier No. NBP271351B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FRA-1/FOSL1 Monoclonal specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FRA-1/FOSL1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI12F9
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, fra-1, FRA1
Gene Symbols FOSL1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human FRA-1/FOSL1 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8061.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.