Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRA-1/FOSL1 Antibody (OTI12F9), PE, Novus Biologicals™
SDP

Catalog No. NB288940 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288940 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288940 Supplier Novus Biologicals Supplier No. NBP271351PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FRA-1/FOSL1 Monoclonal antibody specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen FRA-1/FOSL1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI12F9
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human FOSL1 expression vector
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8061
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.