Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRA10AC1 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB132437 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132437 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132437 Supplier Novus Biologicals Supplier No. NBP306114AF647

Rabbit Polyclonal Antibody

FRA10AC1 Polyclonal specifically detects FRA10AC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FRA10AC1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C10orf4fragile site 10q23.3, chromosome 10 open reading frame 4, F26C11.1-like, FRA10A, fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate1, protein FRA10AC1, rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FRA10AC1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.