Learn More
United Scientific™ Fractional Weight Set
Shop All United Scientific Supplies, Inc Products
Click to view available options
Material:
Steel
Description
- Set includes 1 each of 1, 5, 10, 50, 100, and 500 mg weights, two each of 2, 20, and 200 mg weights, rider, forceps and rectangular plastic box.
- Also available as a set without forceps in a round plastic box.
- Riders are also available separately.
Specifications
Specifications
| Product Type | Weight Set |
| Material | Steel |
| Weight (Metric) | Assorted |
Warning: Cancer and reproductive harm