Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRAT2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001009 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001009 Supplier Novus Biologicals Supplier No. NB1001009
Add to Cart
Add to Cart

Goat Polyclonal Antibody

FRAT2 Polyclonal specifically detects FRAT2 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen FRAT2
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O75474
Gene Alias frequently rearranged in advanced T-cell lymphomas 2FRAT-2, GSK-3 binding protein FRAT2, GSK-3-binding protein FRAT2, MGC10562
Gene Symbols FRAT2
Host Species Goat
Immunogen Peptide with sequence C-GPDRIALQPSGSLL corresponding to C-Terminus according to NP_036215.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23401
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.