Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Antibody (OTI3C7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270769A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270769A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270769A48 Supplier Novus Biologicals Supplier No. NBP270769AF488

Mouse Monoclonal Antibody

Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Western Blot.

Specifications

Antigen Frataxin
Applications Western Blot
Classification Monoclonal
Clone OTI3C7
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Gene Symbols FXN
Host Species Mouse
Immunogen Full length recombinant protein of human Frataxin (NP_000135) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2395.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.