Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Antibody (rFXN/2124), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB379744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379744 Supplier Novus Biologicals Supplier No. NBP308576V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for ELISA

Specifications

Antigen Frataxin
Applications ELISA
Classification Monoclonal
Clone rFXN/2124
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Host Species Mouse
Immunogen Recombinant human Frataxin protein fragment (around aa 57-210) (exact sequence is proprietary) (Uniprot: Q16595)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2395
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.