Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Mouse anti-Human, PE, Clone: 1000000000000, Novus Biologicals™
SDP

Catalog No. NB162190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162190 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162190 Supplier Novus Biologicals Supplier No. NBP259478PE
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Frataxin
Applications Flow Cytometry
Classification Monoclonal
Clone 1000000000000
Conjugate PE
Formulation PBS
Gene Alias CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Host Species Mouse
Immunogen Recombinant human Frataxin (42-210aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2395
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.