Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FRAX 486

Click to view available options
Quantity:
10 mg
Description
FRAX 486 is a potent p21-activated kinase (PAK) inhibitor (IC50 values are 14, 33, 39 and 575 nM for PAK1, PAK2, PAK3 and PAK4 respectively). Blocks and reverses the DISC1 knockdown-induced reduction in dendritic spine size in cortical neurons. Attenuates dendritic spine elimination and enhances spine generation in DISC1 knockdown mice. Ameliorates autism-like behavioral symptoms in fragile X mental retardation 1 (Fmr1) knockout mice. Brain penetrant and orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Product Type | FRAX 486 |
Purity | >98% |
Molecular Weight (g/mol) | 513.39 |
Inhibitors | FRAX 486 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction