Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FRAX 597

Click to view available options
Quantity:
10 mg
50 mg
Description
FRAX 597 is a potent group I PAK inhibitor (IC50 values are 8, 13 and 19 nM for PAK1, 2 and 3, respectively). FRAX 597 exhibits significant inhibition of YES1, RET, CSF1R and TEK at 100 nM, but is inactive against group II PAK isoforms (IC50 >10 μM for PAK4). FRAX 597 inhibits proliferation of pancreatic cancer and schwannoma cells in vitro and exhibits antitumor effects in mice.
Specifications
Specifications
| Chemical Name or Material | FRAX 597 |
| CAS | 1286739-19-2 |
| Synonym | 6-[2-Chloro-4-(5-thiazolyl)phenyl]-8-ethyl-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7-(8H)-one |
| Quantity | 50 mg |
| Solubility | Soluble to 20mM in DMSO |
| Target | PAK Inhibitors |
| Molecular Formula | C29H28ClN7OS |
| Formula Weight | 558.1 |
| Physical Form | Solid |
| Color | Yellow |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction