Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FREM1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7205275 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8478 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8478 Supplier Proteintech Group Inc Supplier No. 130861AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes isoform 1 (244 kDa), isoform 2 (80 kDa) and isoform 3 (43 kDa).

FREM1 is a member of the FRAS1-related extracellular matrix protein family and is thought to play a role in craniofacial and renal development. FREM1 functions as an extracellular matrix protein that is essential for epidermal adhesion during embryogenesis and may also participate in epidermal differentiation. It is recognized by cells in the embryonic skin and hair follicles through different members of the integrin family. Deficiency in the Fras1/Frem genes gives rise to the bleb phenotype, which is equivalent to the human hereditary disorder Fraser syndrome.

Specifications

Antigen FREM1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FREM1
Gene Accession No. Q5H8C1
Gene Alias C9orf143, C9orf145, C9orf154, FREM1, Protein QBRICK, QBRICK, RP11 439N12.3
Gene Symbols FREM1
Host Species Rabbit
Immunogen FREM1 Fusion Protein Ag3617
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.