Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frequenin Antibody (11B8), PerCP, Novus Biologicals™
SDP

Catalog No. NB162234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162234 Supplier Novus Biologicals Supplier No. NBP259480PCP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frequenin Monoclonal antibody specifically detects Frequenin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Frequenin
Applications Flow Cytometry
Classification Monoclonal
Clone 11B8
Conjugate PerCP
Formulation PBS
Gene Alias DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1
Host Species Mouse
Immunogen Recombinant human Frequenin (1-190aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23413
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.