Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-10 Antibody (2678A), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB303985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303985 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303985 Supplier Novus Biologicals Supplier No. FAB34581MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Frizzled-10 Monoclonal specifically detects Frizzled-10 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry.

Specifications

Antigen Frizzled-10
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry
Classification Monoclonal
Clone 2678A
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human Frizzled-10, Ile21-Gly161, Accession # Q6NSL8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 11211
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.