Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223659SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223659SS 0.025 mL
NBP223659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223659SS Supplier Novus Biologicals Supplier No. NBP223659SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Frizzled-10 Polyclonal specifically detects Frizzled-10 in Human, Mouse, Primate samples. It is validated for Western Blot.

Specifications

Antigen Frizzled-10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1: 1000
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Alias CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10
Gene Symbols FZD10
Host Species Rabbit
Immunogen A portion of amino acid 150-200 of human Frizzled-10 was used as the immunogen
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 11211
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and monkey and 88% homologous in rat.
Target Species Human, Mouse, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.