Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095477 100 μL
NB095478 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095477 Supplier Novus Biologicals Supplier No. NBP303466100UL

Rabbit Polyclonal Antibody

Frizzled-10 Polyclonal specifically detects Frizzled-10 in Human samples. It is validated for Western Blot.

Specifications

Antigen Frizzled-10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 135-230 of human Frizzled-10 (NP_009128.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 11211
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.