Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120920 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120920 Supplier Novus Biologicals Supplier No. NBP120920
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Frizzled-2 Polyclonal specifically detects Frizzled-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Frizzled-2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03-1 ug/ml, Immunohistochemistry 2 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 2 - 5 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q14332
Gene Alias frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Fz2, Fz-2, fzE2, hFz2
Gene Symbols FZD2
Host Species Goat
Immunogen Peptide with sequence RYATLEHPFHCPR corresponding to internal region according to NP_001457.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2535
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.