Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-3 Antibody (169310) [PerCP], Novus Biologicals™
SDP

Catalog No. NB402696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402696 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402696 Supplier Novus Biologicals Supplier No. FAB1001C

Rat Monoclonal Antibody

Frizzled-3 Monoclonal antibody specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen Frizzled-3
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 169310
Conjugate PerCP
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Fz-3, hFz3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3, Met1-Val157, Accession # Q61086
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7976
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.