Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-4 Antibody (145901), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349446 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349446 Supplier Novus Biologicals Supplier No. FAB194MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Frizzled-4 Monoclonal specifically detects Frizzled-4 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen Frizzled-4
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 145901
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD344, CD344 antigen, EVR1, exudative vitreoretinopathy 1, FEVR, frizzled (Drosophila) homolog 4, frizzled homolog 4 (Drosophila), frizzled-4, Fz-4, FZD4S, FzE4, GPCR, hFz4, MGC34390, WNT receptor frizzled-4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-4, Phe37-Glu180, Accession # Q61088
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8322
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.