Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-6 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB172071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172071 Supplier Novus Biologicals Supplier No. NBP318335AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Frizzled-6 Polyclonal antibody specifically detects Frizzled-6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Frizzled-6
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias frizzled (Drosophila) homolog 6, frizzled homolog 6 (Drosophila), frizzled-6, fz-6, FZ6, HFZ6, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids in the central domain of human Frizzled-6. The immunogen is located within 50-550 amino acids of Frizzled-6.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8323
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.