Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRMD5 Antibody (CL9503), Novus Biologicals™
SDP

Catalog No. NB036528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036528 Supplier Novus Biologicals Supplier No. NBP288935
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FRMD5 Monoclonal specifically detects FRMD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FRMD5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL9503
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, containing 40% glycerol
Gene Alias FERM domain containing 5, FERM domain-containing protein 5, FLJ41022, MGC14161
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SCPSITHGPRLSSVPRTRRRAVHISIMEGLESLRDSAHSTPVRSTSHGDTFLPHVRSSRTDSNERVAVIADEAYSPADSVLPTPVAEHSLELMLLSRQINGATCSIEEEKESEAST
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84978
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.