Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRS2 Antibody (T5), Novus Biologicals™
SDP

Catalog No. NB019500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019500 0.05 mg
NBP229886 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019500 Supplier Novus Biologicals Supplier No. NBP2298860.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FRS2 Monoclonal specifically detects FRS2 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen FRS2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone T5
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation 70mM Tris-Glycine (pH 7.4), 105mM NaCl and 30% Glycerol with Sodium Azide
Gene Accession No. Q8WU20
Gene Alias FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1
Gene Symbols FRS2
Host Species Mouse
Immunogen Peptide corresponding to residues 283-297 of human FRS2/SNT-1.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10818
Test Specificity FRS2/SNT-1 in unstimulated cells and phosphorylated FRS2/SNT-1 in NGF-stimulated cells non-phosphorylated(68kDa) and phosphorylated(83kDa) weight may be detected.
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.