Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRS2 Antibody (T5), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $731.50
Specifications
Antigen | FRS2 |
---|---|
Clone | T5 |
Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
FRS2 Monoclonal specifically detects FRS2 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
FRS2 | |
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Mouse | |
Cancer, Signal Transduction | |
Q8WU20 | |
10818 | |
Peptide corresponding to residues 283-297 of human FRS2/SNT-1. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
T5 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Rat | |
FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
FRS2 | |
IgG | |
Affinity Purified | |
FRS2/SNT-1 in unstimulated cells and phosphorylated FRS2/SNT-1 in NGF-stimulated cells non-phosphorylated(68kDa) and phosphorylated(83kDa) weight may be detected. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title