Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fructosyl amino acid oxidase Antibody, Novus Biologicals™
SDP

Catalog No. NBP17778201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17778201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17778201 Supplier Novus Biologicals Supplier No. NBP1777820.1MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Fructosyl amino acid oxidase Polyclonal specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Fructosyl amino acid oxidase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q8RIU8
Gene Symbols faoxC
Host Species Sheep
Immunogen Fructosyl-Amino Acid Oxidase [from Corynebacterium sp. Expressed in E.coli]
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.